Anti-IgG (H+L) Goat Polyclonal Antibody

Leverandør: Agrisera
AS10773
AGRIAS10773EA 2650 NOK
AGRIAS10773
Anti-IgG (H+L) Goat Polyclonal Antibody
Antistoffer
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Human, Mouse, Rabbit
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR