Anti-IgG (H+L) Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Leverandør: Agrisera
AS10957
AGRIAS10957EA 2250 NOK
AGRIAS10957
Anti-IgG (H+L) Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antistoffer
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rabbit
Bestill nå

SPESIFIKASJONER

Antigensymbol IgG (H+L)
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering TRITC (Tetramethylrhodamine Isothiocyanate)
Reaktivitet Rabbit,Human,Mouse
Vert Rabbit
ELISA Yes
ImmunoKjemi Yes
Flowcytometri Yes
Immunogen Purified Mouse IgG, whole molecule
Rensing purified
Kyssabsorpsjon No
Lagringstemp +2°C to +8 °C
Frakttemp. Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR