Anti-PIN2 Rabbit Polyclonal Antibody (KLH)

Leverandør: Agrisera
AS204521
AGRIAS204521EA 3850 NOK
AGRIAS204521
Anti-PIN2 Rabbit Polyclonal Antibody (KLH)
Antistoffer
Recommended dilution: 1 : 200 (IL)

Type:
Antigen: PIN2
Clonality: Polyclonal
Clone:
Conjugation: KLH
Epitope:
Host: Rabbit
Isotype:
Reactivity: Arabidopsis thaliana
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR