Precision thermometer
Leverandør: Amarell
M242362-TOL
AMARM242362-TOLEA 1150 NOK
AMARM242362-TOL
Precision thermometer
Termometere Analogue Thermometers
This thermometer is suitable for government verification.
  • Solid stem
  • Top round finish
  • White backed
Toluene filling.
Sertifiseringer: Conforms to DIN 12785.