Bi-metallic dial thermometer
Leverandør: Amarell
T110410
AMART110410EA 0 NOK
AMART110410
Bi-metallic dial thermometer
Termometere Analogue Thermometers
Bimetal dial thermometers made of brass. The scale cover made of glass is used in heating systems and indutrial plants.
  • Axial stem
  • Thread: R ½"
  • Quick and reliable temperature control