Stool and sputum cups
Leverandør: Anicrin
C018S110
ANICC018S110EA 2425 NOK
ANICC018S110
Stool and sputum cups
Prøvecontainere Beholdere, generelt bruk
PS, translucent, with white PE stopper
  • Small spoon attached to stopper
  • Good thermal resistance, good chemical resistance
  • Universal application, ideal for single use
Straight-walled containers for sample collection, storage and transport.