Anti-KLH Mouse Monoclonal Antibody [clone: KT85]

Leverandør: ANTIBODIES.COM
A131-100 A131-200
ANTIA131-100EA 5475 NOK
ANTIA131-100 ANTIA131-200
Anti-KLH Mouse Monoclonal Antibody [clone: KT85]
Antistoffer
Mouse monoclonal (KT85) antibody to KLH for isotype control.

  • Validated applications: Isotype control
  • Host: Mouse
  • Clonality: Monoclonal
  • Isotype: IgG3
  • Conjugate: Unconjugated

Type: Primary
Antigen: KLH
Clonality: Monoclonal
Clone: KT85
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
New Product
Bestill nå

SPESIFIKASJONER

Antigensymbol KLH
Antigen-navn Hemocyanin 2 c chain
Environmentally Preferable
Antigen synonymer Hemocyanin 2c chain|KLH B|KLH A|KLH|KLH2 c|Hemocyanin B chain|Hemocyanin A chain|KLH2c|Keyhole limpet hemocyanin
Antistofftype Primary
Klonalitet Monoclonal
Konjugering Unconjugated
Klon KT85
Reaktivitet
Vert Mouse
Isotype IgG3
Immunogen Keyhole limpet hemocyanin
Skjema Liquid
Rensing IgG is purified from culture supernatant through a Protein A column
Konsentrasjon 1 mg/ml
Lagringsbuffer Supplied in phosphate buffered saline with 0,09% sodium azide
Lagringstemp Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
Frakttemp. Shipped on blue ice at 4 °C
Uttestet applikasjon Isotype control

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR