Anti-KLH Mouse Monoclonal Antibody [clone: KT83]

Leverandør: ANTIBODIES.COM
A134-100 A134-200
ANTIA134-100EA 5475 NOK
ANTIA134-100 ANTIA134-200
Anti-KLH Mouse Monoclonal Antibody [clone: KT83]
Antistoffer
Mouse monoclonal (KT83) antibody to KLH for isotype control.

  • Validated applications: Isotype control
  • Host: Mouse
  • Clonality: Monoclonal
  • Isotype: IgG2a
  • Conjugate: Unconjugated

Type: Primary
Antigen: KLH
Clonality: Monoclonal
Clone: KT83
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR