Anti-KLH Mouse Monoclonal Antibody [clone: KT86]

Leverandør: ANTIBODIES.COM
A135-100
ANTIA135-100EA 5475 NOK
ANTIA135-100
Anti-KLH Mouse Monoclonal Antibody [clone: KT86]
Antistoffer
Mouse monoclonal (KT86 ) antibody to KLH for Isotype Control.

  • Validated applications: Isotype Control

Type: Primary
Antigen: KLH
Clonality: Monoclonal
Clone: KT86
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgA
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR