Anti-Glyoxalase 1 Mouse Monoclonal Antibody [clone: CPTC-GLO1-1]

Leverandør: ANTIBODIES.COM
A248739-100
ANTIA248739-100EA 6400 NOK
ANTIA248739-100
Anti-Glyoxalase 1 Mouse Monoclonal Antibody [clone: CPTC-GLO1-1]
Antistoffer
Mouse monoclonal [CPTC-GLO1-1] antibody to Glyoxalase 1 for WB and IHC-P with samples derived from human.

  • Validated applications: WB, IHC-P
  • Recommended dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Glyoxalase 1
Clonality: Monoclonal
Clone: CPTC-GLO1-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR