Anti-NCAM1 Mouse Monoclonal Antibody [clone: NCAM1/795]

Leverandør: ANTIBODIES.COM
A249480-100
ANTIA249480-100EA 6400 NOK
ANTIA249480-100
Anti-NCAM1 Mouse Monoclonal Antibody [clone: NCAM1/795]
Antistoffer
Mouse monoclonal [NCAM1/795] antibody to NCAM1 for flow cytometry, IF, WB and IHC-P with samples derived from human and rat.

  • Validated applications: Flow cytometry, IF, WB, IHC-P
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: NCAM1
Clonality: Monoclonal
Clone: NCAM1/795
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rat
New Product
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR