Anti-Spermidine Mouse Monoclonal Antibody [clone: CPTC-SAT1-3]

Leverandør: ANTIBODIES.COM
A253125-100
ANTIA253125-100EA 6400 NOK
ANTIA253125-100
Anti-Spermidine Mouse Monoclonal Antibody [clone: CPTC-SAT1-3]
Antistoffer
Mouse monoclonal [CPTC-SAT1-3] antibody to Spermidine for ELISA with samples derived from Human.

  • Validated applications: ELISA

Type: Primary
Antigen: Spermidine
Clonality: Monoclonal
Clone: CPTC-SAT1-3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
New Product
Bestill nå

SPESIFIKASJONER

Antigensymbol Spermidine
Environmentally Preferable
Antistofftype Primary
Klonalitet Monoclonal
Konjugering Unconjugated
Klon CPTC-SAT1-3
Reaktivitet Human
Vert Mouse
Gen-ID UniprotID# P21673
Isotype IgG2a kappa
ELISA Yes
Immunogen Recombinant full-length human SAT1 protein
Skjema Liquid
Molekylvekt 20 kDa
Rensing Protein A/G chromatography
Konsentrasjon 1 mg/ml
Lagringsbuffer Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
Lagringstemp Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
Frakttemp. Shipped on blue ice at 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR