Anti-beta III Spectrin Mouse Monoclonal Antibody [clone: RG/26]

Leverandør: ANTIBODIES.COM
A253222-100
ANTIA253222-100EA 6400 NOK
ANTIA253222-100
Anti-beta III Spectrin Mouse Monoclonal Antibody [clone: RG/26]
Antistoffer
Mouse monoclonal [RG/26] antibody to beta III Spectrin for ELISA, WB, IF and IHC-P with samples derived from Human, Mouse and Rat.

  • Validated applications: ELISA, WB, IF, IHC-P
  • Recommended dilutions: WB: 1 to 2 µg/ml, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: beta III Spectrin
Clonality: Monoclonal
Clone: RG/26
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR