Mouse Anti-KLH IgG ELISA Kit
Leverandør: ANTIBODIES.COM
A303554-96
ANTIA303554-96EA 7750 NOK
ANTIA303554-96
Mouse Anti-KLH IgG ELISA Kit
Assay ELISAs
Mouse Anti-KLH IgG ELISA kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of mouse Anti-KLH IgG in serum, plasma, tissue homogenates, and other biological fluids.