Anti-PSD95 Mouse Monoclonal Antibody [Clone: 7E3]

Leverandør: ANTIBODIES.COM
A304702-100
ANTIA304702-100EA 5550 NOK
ANTIA304702-100
Anti-PSD95 Mouse Monoclonal Antibody [Clone: 7E3]
Antistoffer
Mouse monoclonal [7E3] antibody to PSD95 for WB, IHC, ICC/IF and antibody microarray with samples derived from human, mouse, rat and bovine.

  • Validated applications: WB, IHC, ICC/IF, Antibody microarray
  • Recommended dilutions: WB: 1:1000, IHC: 1:1000, ICC/IF: 1:100

Type: Primary
Antigen: PSD95
Clonality: Monoclonal
Clone: 7E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Bovine
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR