Anti-PSD95 Mouse Monoclonal Antibody [clone: 6G6]

Leverandør: ANTIBODIES.COM
A304703-100
ANTIA304703-100EA 5550 NOK
ANTIA304703-100
Anti-PSD95 Mouse Monoclonal Antibody [clone: 6G6]
Antistoffer
Mouse monoclonal [6G6] antibody to PSD95 for WB, IHC, ICC/IF and antibody microarray with samples derived from human, mouse, rat and bovine.

  • Validated applications: WB, IHC, ICC/IF, Antibody microarray
  • Recommended dilutions: WB: 1:250, IHC: 1:1000, ICC/IF: 1:100

Type: Primary
Antigen: PSD95
Clonality: Monoclonal
Clone: 6G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human;Mouse;Rat;Bovine
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR