Anti-HCN1 Mouse Monoclonal Antibody [Clone: S70]

Leverandør: ANTIBODIES.COM
A304768-100
ANTIA304768-100EA 5850 NOK
ANTIA304768-100
Anti-HCN1 Mouse Monoclonal Antibody [Clone: S70]
Antistoffer
Mouse monoclonal [S70] antibody to HCN1 for WB, IHC, IP and antibody microarray with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC, IP, Antibody microarray
  • Recommended dilutions: WB: 1:1000, IHC: 1:1000, ICC/IF: 1:100

Type: Primary
Antigen: HCN1
Clonality: Monoclonal
Clone: S70
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR