Anti-HSP70 Mouse Monoclonal Antibody [clone: C92F3A-5]

Leverandør: ANTIBODIES.COM
A304773-200
ANTIA304773-200EA 6275 NOK
ANTIA304773-200
Anti-HSP70 Mouse Monoclonal Antibody [clone: C92F3A-5]
Antistoffer
Mouse monoclonal [C92F3A-5] antibody to HSP70 for WB, IHC, ICC/IF, ELISA, Flow cytometry, FACS, IEM, Block/Inhibit and antibody microarray with samples derived from human, mouse, rat, bovine, C. elegans, canine, chicken, drosophila, carp, guinea pig, hamster, monkey, porcine, rabbit and sheep.

  • Validated applications: WB, IHC, ICC/IF, ELISA, Flow cytometry, FACS, IEM, Block/Inhibit, Antibody microarray
  • Recommended dilutions: WB: 1:1000, IHC: 1:10000, ICC/IF: 1:1000, FACS: 1:1000

Type: Primary
Antigen: HSP70
Clonality: Monoclonal
Clone: C92F3A-5
Conjugation:More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR