Anti-HSC70 Mouse Monoclonal Antibody [Clone: 1F2-H5]

Leverandør: ANTIBODIES.COM
A304778-200
ANTIA304778-200EA 6450 NOK
ANTIA304778-200
Anti-HSC70 Mouse Monoclonal Antibody [Clone: 1F2-H5]
Antistoffer
Mouse monoclonal [1F2-H5] antibody to HSC70 for WB, IHC, ICC/IF, IP, ELISA, PLA, PBA and antibody microarray with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC, ICC/IF, IP, ELISA, PLA, PBA, Antibody microarray
  • Recommended dilutions: WB: 1:1000, ICC/IF: 1:100

Type: Primary
Antigen: HSC70
Clonality: Monoclonal
Clone: 1F2-H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR