Anti-KCNT1/SLACK Mouse Monoclonal Antibody [clone: N3/26]

Leverandør: ANTIBODIES.COM
A304785-100
ANTIA304785-100EA 5850 NOK
ANTIA304785-100
Anti-KCNT1/SLACK Mouse Monoclonal Antibody [clone: N3/26]
Antistoffer
Mouse monoclonal [N3/26] antibody to KCNT1/SLACK for WB, IHC, ICC/IF and antibody microarray with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC, ICC/IF, Antibody microarray
  • Recommended dilutions: WB: 1:1000, IHC: 1:1000, ICC/IF: 1:100

Type: Primary
Antigen: KCNT1/SLACK
Clonality: Monoclonal
Clone: N3/26
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR