Anti-Kir2.1 Mouse Monoclonal Antibody [Clone: S112]

Leverandør: ANTIBODIES.COM
A304786-100
ANTIA304786-100EA 5850 NOK
ANTIA304786-100
Anti-Kir2.1 Mouse Monoclonal Antibody [Clone: S112]
Antistoffer
Mouse monoclonal [S112] antibody to Kir2.1 for WB, IHC, ICC/IF and antibody microarray with samples derived from human, mouse, rat and monkey.

  • Validated applications: WB, IHC, ICC/IF, Antibody microarray
  • Recommended dilutions: WB: 1:1000, IHC: 1:1000, ICC/IF: 1:100

Type: Primary
Antigen: Kir2.1
Clonality: Monoclonal
Clone: S112
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Monkey
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR