Anti-DMPO Mouse Monoclonal Antibody [clone: N1664A]

Leverandør: ANTIBODIES.COM
A304793-100
ANTIA304793-100EA 6025 NOK
ANTIA304793-100
Anti-DMPO Mouse Monoclonal Antibody [clone: N1664A]
Antistoffer
Mouse monoclonal [N1664A] antibody to DMPO for WB, IHC, ICC/IF, IP, ELISA and antibody microarray with samples derived from species independent.

  • Validated applications: WB, IHC, ICC/IF, IP, ELISA, Antibody microarray
  • Recommended dilutions: WB: 1:1000, ICC/IF: 1:100, ELISA: 1:100, IP: 25 µg

Type: Primary
Antigen: DMPO
Clonality: Monoclonal
Clone: N1664A
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Species Independent
New Product
Bestill nå

SPESIFIKASJONER

Antigensymbol DMPO
Environmentally Preferable
Antistofftype Primary
Klonalitet Monoclonal
Konjugering Unconjugated
Klon N1664A
Reaktivitet Species independent
Vert Mouse
Isotype IgG1
Western blot Yes
ELISA Yes
ImmunoKjemi Yes
Immunutfelling Yes
Immunogen 5,5 - dimethyl - 2 - (8 - octanoic acid) - 1 - pyrrolone - N - oxide conjugated to Ovalbumin.
Skjema Liquid
Rensing Protein G purification
Konsentrasjon 1 mg/ml
Lagringsbuffer Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol and 0,09% sodium azide
Lagringstemp Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
Frakttemp. Shipped on blue ice at 4 °C
Uttestet applikasjon IHC, ICC/IF

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR