Anti-Nitrotyrosine Mouse Monoclonal Antibody [Clone: 39B6]

Leverandør: ANTIBODIES.COM
A304794-100
ANTIA304794-100EA 6025 NOK
ANTIA304794-100
Anti-Nitrotyrosine Mouse Monoclonal Antibody [Clone: 39B6]
Antistoffer
Mouse monoclonal [39B6] antibody to Nitrotyrosine for WB, IHC, ICC/IF, IP, ELISA, Flow cytometry and antibody microarray with samples derived from species independent.

  • Validated applications: WB, IHC, ICC/IF, IP, ELISA, Flow cytometry, Antibody microarray
  • Recommended dilutions: WB: 1:1400, IHC: 1:100

Type: Primary
Antigen: Nitrotyrosine
Clonality: Monoclonal
Clone: 39B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Species Independent
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR