Anti-HCN4 Mouse Monoclonal Antibody [Clone: N114/10]

Leverandør: ANTIBODIES.COM
A304800-100
ANTIA304800-100EA 5850 NOK
ANTIA304800-100
Anti-HCN4 Mouse Monoclonal Antibody [Clone: N114/10]
Antistoffer
Mouse monoclonal [N114/10] antibody to HCN4 for WB, IHC, ICC/IF, antibody microarray and FACS with samples derived from human, mouse, rat and rabbit.

  • Validated applications: WB, IHC, ICC/IF, Antibody microarray, FACS
  • Recommended dilutions: WB: 1:1000, IHC: 1:1000, ICC/IF: 1:100

Type: Primary
Antigen: HCN4
Clonality: Monoclonal
Clone: N114/10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Rabbit
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR