Anti-Nav1.7 Mouse Monoclonal Antibody [clone: N68/6]

Leverandør: ANTIBODIES.COM
A304801-100
ANTIA304801-100EA 5850 NOK
ANTIA304801-100
Anti-Nav1.7 Mouse Monoclonal Antibody [clone: N68/6]
Antistoffer
Mouse monoclonal [N68/6] antibody to Nav1.7 for WB, IHC, ICC/IF, IP and antibody microarray with samples derived from human, mouse, rat and hamster.

  • Validated applications: WB, IHC, ICC/IF, IP, Antibody microarray
  • Recommended dilutions: WB: 1:1000, IHC: 1:1000, ICC/IF: 1:100

Type: Primary
Antigen: Nav1.7
Clonality: Monoclonal
Clone: N68/6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat;Hamster
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR