Anti-Nav1.8 Mouse Monoclonal Antibody [Clone: S134]

Leverandør: ANTIBODIES.COM
A304802-100
ANTIA304802-100EA 5850 NOK
ANTIA304802-100
Anti-Nav1.8 Mouse Monoclonal Antibody [Clone: S134]
Antistoffer
Mouse monoclonal [S134] antibody to Nav1.8 for WB, IHC, ICC/IF and antibody microarray with samples derived from human, mouse, rat and monkey.

  • Validated applications: WB, IHC, ICC/IF, Antibody microarray
  • Recommended dilutions: WB: 1:1000, IHC: 1:1000, ICC/IF: 1:100

Type: Primary
Antigen: Nav1.8
Clonality: Monoclonal
Clone: S134
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat, Monkey
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR