Anti-TSP23 Mouse Monoclonal Antibody [clone: Tsp232A]

Leverandør: ANTIBODIES.COM
A304805-100
ANTIA304805-100EA 5550 NOK
ANTIA304805-100
Anti-TSP23 Mouse Monoclonal Antibody [clone: Tsp232A]
Antistoffer
Mouse monoclonal [Tsp232A] antibody to TSP23 for WB, IHC and antibody microarray with samples derived from human and mouse.

  • Validated applications: WB, IHC, Antibody microarray
  • Recommended dilutions: WB: 1:1000, IHC: 1:1000

Type: Primary
Antigen: TSP23
Clonality: Monoclonal
Clone: Tsp232A
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR