Anti-SHANK1 Mouse Monoclonal Antibody [clone: N22/21]

Leverandør: ANTIBODIES.COM
A304815-100
ANTIA304815-100EA 5850 NOK
ANTIA304815-100
Anti-SHANK1 Mouse Monoclonal Antibody [clone: N22/21]
Antistoffer
Mouse monoclonal [N22/21] antibody to SHANK1 for WB, IHC and antibody microarray with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC, Antibody microarray
  • Recommended dilutions: WB: 1:1000, IHC: 1:1000, ICC/IF: 1:100

Type: Primary
Antigen: SHANK1
Clonality: Monoclonal
Clone: N22/21
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR