Anti-p38 alpha/MAPK14 Mouse Monoclonal Antibody [Clone: 9F12]

Leverandør: ANTIBODIES.COM
A304996-100
ANTIA304996-100EA 4875 NOK
ANTIA304996-100
Anti-p38 alpha/MAPK14 Mouse Monoclonal Antibody [Clone: 9F12]
Antistoffer
Mouse monoclonal [9F12] antibody to p38 alpha/MAPK14 for WB, IHC, IP, ELISA and antibody microarray with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC, IP, ELISA, Antibody microarray
  • Recommended dilutions: WB: 1:1000, IHC: 1:1000

Type: Primary
Antigen: p38 alpha/MAPK14
Clonality: Monoclonal
Clone: 9F12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
New Product
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR