Anti-GABRD Mouse Monoclonal Antibody [Clone: N151/3]

Leverandør: ANTIBODIES.COM
A305006-100
ANTIA305006-100EA 5850 NOK
ANTIA305006-100
Anti-GABRD Mouse Monoclonal Antibody [Clone: N151/3]
Antistoffer
Mouse monoclonal [N151/3] antibody to GABRD for WB, ICC/IF and antibody microarray with samples derived from human, mouse and rat.

  • Validated applications: WB, ICC/IF, Antibody microarray
  • Recommended dilutions: WB: 1:1000, IHC: 1:1000, ICC/IF: 1:100

Type: Primary
Antigen: GABRD
Clonality: Monoclonal
Clone: N151/3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
New Product
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR