Anti-TRPV3 Mouse Monoclonal Antibody [clone: N15/39]

Leverandør: ANTIBODIES.COM
A305015-100
ANTIA305015-100EA 5850 NOK
ANTIA305015-100
Anti-TRPV3 Mouse Monoclonal Antibody [clone: N15/39]
Antistoffer
Mouse monoclonal [N15/39] antibody to TRPV3 for WB, IHC, ICC/IF, IP and antibody microarray with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC, ICC/IF, IP, Antibody microarray
  • Recommended dilutions: WB: 1:1000, IHC: 1:1000, ICC/IF: 1:100

Type: Primary
Antigen: TRPV3
Clonality: Monoclonal
Clone: N15/39
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human;Mouse;Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR