Anti-Sodium Iodide Symporter Mouse Monoclonal Antibody [clone: 14F]

Leverandør: ANTIBODIES.COM
A305039-100
ANTIA305039-100EA 5850 NOK
ANTIA305039-100
Anti-Sodium Iodide Symporter Mouse Monoclonal Antibody [clone: 14F]
Antistoffer
Mouse monoclonal [14F] antibody to Sodium Iodide Symporter for WB, IHC and antibody microarray with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC, Antibody microarray
  • Recommended dilutions: WB: 1:1000, IHC: 1:1000

Type: Primary
Antigen: Sodium Iodide Symporter
Clonality: Monoclonal
Clone: 14F
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR