Anti-Grp75/MOT Rabbit Monoclonal Antibody [clone: ARC0566]

Leverandør: ANTIBODIES.COM
A305679-100
ANTIA305679-100EA 6450 NOK
ANTIA305679-100
Anti-Grp75/MOT Rabbit Monoclonal Antibody [clone: ARC0566]
Antistoffer
Rabbit monoclonal [ARC0566] antibody to Grp75/MOT for WB with samples derived from human, mouse and rat.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:500 to 1:2000

Type: Primary
Antigen: Grp75/MOT
Clonality: Monoclonal
Clone: ARC0566
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR