Anti-PITX2/RGS Rabbit Monoclonal Antibody [clone: ARC56103]

Leverandør: ANTIBODIES.COM
A307302-100
ANTIA307302-100EA 6450 NOK
ANTIA307302-100
Anti-PITX2/RGS Rabbit Monoclonal Antibody [clone: ARC56103]
Antistoffer
Rabbit monoclonal [ARC56103] antibody to PITX2/RGS for WB with samples derived from Human.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:1000 to 1:5000

Type: Primary
Antigen: PITX2 / RGS
Clonality: Monoclonal
Clone: ARC56103
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Bestill nå

SPESIFIKASJONER

Antigensymbol PITX2 / RGS
Environmentally Preferable
Antistofftype Primary
Klonalitet Monoclonal
Konjugering Unconjugated
Klon ARC56103
Reaktivitet Human
Vert Rabbit
Gen-ID UniprotID# Q99697
Isotype IgG
Western blot Yes
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Pituitary homeobox 2 (PITX2) (NP_700475.1).
Sekvens METNCRKLVSACVQLGVQPAAVECLFSKDSEIKKVEFTDSPESRKEAASSKFFPRQHPGANEKDKSQQGKNEDVGAEDPSKKKRQRRQRTHFTSQQLQEL
Skjema Liquid
Molekylvekt 37 kDa
Rensing Affinity purification
Lagringsbuffer Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,05% Proclin 300
Lagringstemp Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
Frakttemp. Shipped on blue ice at 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR