Anti-Puromycin Rabbit Polyclonal Antibody

Leverandør: ANTIBODIES.COM
A309724-100
ANTIA309724-100EA 5100 NOK
ANTIA309724-100
Anti-Puromycin Rabbit Polyclonal Antibody
Antistoffer
Rabbit polyclonal antibody to Puromycin for WB with samples derived from human and mouse.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:1,000 to 1:5,000

Type: Primary
Antigen: Puromycin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR