Human PITX2/RGS ELISA Kit

Leverandør: ANTIBODIES.COM

A312882-96
ANTIA312882-96EA 6700 NOK
ANTIA312882-96
Human PITX2/RGS ELISA Kit
Assay ELISAs

Human PITX2/RGS ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human PITX2/RGS in serum, plasma, and other biological fluids.


  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 100 to 1600 ng/L
  • Sensitivity: 7,28 ng/L
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%


Human PITX2/RGS ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human PITX2/RGS in serum, plasma or other biological fluids. An antibody specific for PITX2/RGS has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the PITX2/RGS present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-PITX2/RGS antibody, which also binds the PITX2/RGS present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-PITX2/RGS antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-PITX2/RGS antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of PITX2/RGS captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of PITX2/RGS in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.


Bestillingsinformasjon: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR