Canine Anti-KLH IgM ELISA Kit
Leverandør: ANTIBODIES.COM
A319512-96
ANTIA319512-96EA 9725 NOK
ANTIA319512-96
Canine Anti-KLH IgM ELISA Kit
Assay ELISAs
Canine Anti-KLH IgM ELISA kit is a capture Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of canine Anti-KLH IgM in serum, plasma, tissue homogenates, and other biological fluids.
  • Assay Precision: Intra-assay CV: <8%, Inter-assay CV: <10%
  • Assay Time: 4 hours
NB: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.