Canine Anti-KLH IgG ELISA Kit

Leverandør: ANTIBODIES.COM

A319521-96
ANTIA319521-96EA 9725 NOK
ANTIA319521-96
Canine Anti-KLH IgG ELISA Kit
Assay ELISAs

Canine Anti-KLH IgG ELISA kit is an indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of canine Anti-KLH IgG in serum, plasma, tissue homogenates, and other biological fluids.


  • Assay Precision: Intra-assay CV: <8%, Inter-assay CV: <10%
  • Assay Time: 4 hours


NB: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.

New Product

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Mål Anti-KLH IgG
Primary antibody reactivity Dog
Analysetype Indirect
Vert
Format Pre-coated
Analysens varighet Multiple steps
Sensitivitet 0,938 ng/ml
Påvisningsspekter 1,563 - 100 ng/ml
Påvisningsmetode Colorimetric
Type prøve Serum, plasma, tissue homogenates, and other biological fluids
Time to Results 4 hours
Regulatorisk status RUO
Lagringstemp 4 °C
Holdbarhet Store for 6 months at 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR