Rat JAK3 ELISA Kit

Leverandør: ANTIBODIES.COM

A80052-96
ANTIA80052-96EA 7375 NOK
ANTIA80052-96
Rat JAK3 ELISA Kit
Assay ELISAs

Rat JAK3 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat JAK3 in serum, plasma, tissue homogenates, and other biological fluids.


  • Ready to use ELISA Kit
  • Detection range: 0,781 to 50 ng/ml
  • Sensitivity: 0,469 ng/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%


Rat JAK3 ELISA Kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat JAK3 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for JAK3 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the JAK3 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-JAK3 Antibody, which binds the captured JAK3 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of JAK3 captured in each well. The concentration of JAK3 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.


Bestillingsinformasjon: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR