Anti-APPL1 Goat Polyclonal Antibody

Leverandør: ANTIBODIES.COM
A84292-100
ANTIA84292-100EA 5850 NOK
ANTIA84292-100
Anti-APPL1 Goat Polyclonal Antibody
Antistoffer
Goat polyclonal antibody to APPL1 for ELISA and WB with samples derived from human and rat.

  • Validated applications: ELISA, W

Type: Primary
Antigen: APPL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR