Anti-CTAG1B Goat Polyclonal Antibody

Leverandør: ANTIBODIES.COM
A84635-100
ANTIA84635-100EA 5850 NOK
ANTIA84635-100
Anti-CTAG1B Goat Polyclonal Antibody
Antistoffer
Goat polyclonal antibody to CTAG1B for ELISA, IF, flow cytometry and Microarray with samples derived from human.

  • Validated applications: ELISA, IF, Flow cytometry, Microarra

Type: Primary
Antigen: CTAG1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
New Product
Bestill nå

SPESIFIKASJONER

Antigensymbol CTAG1B
Environmentally Preferable
Antigen-navn Autoimmunogenic cancer/testis antigen NY ESO 1
Antigen synonymer Autoimmunogenic cancer/testis antigen NY-ESO-1|Cancer/testis antigen 1B|CT6.1|CTAG 1B|Cancer antigen 3|CTAG|CTAG 1|Cancer/testis antigen 6.1|Cancer/testis antigen 1
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human
Vert Goat
Isotype IgG
ELISA Yes
ImmunoFluorescens Yes
Flowcytometri Yes
Immunogen Synthetic peptide corresponding to Human CTAG1B (internal region)
Sekvens C-AELARRSLAQDAP
Skjema Liquid
Rensing Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
Konsentrasjon 100 µg at 0,5 mg/ml
Lagringsbuffer Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
Lagringstemp Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
Frakttemp. Shipped on blue ice at 4 °C
Uttestet applikasjon Microarray

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR