Antipain dihydrochloride
Leverandør: Apollo Scientific
BIA0201-10MG BIA0201-5MG
APOSBIA0201-10MGEA 805 NOK
APOSBIA0201-10MG APOSBIA0201-5MG
Antipain dihydrochloride
Proteiner
Serine and cysteine protease inhibitor
Bestill nå
SPESIFIKASJONER
Protein/peptide name Antipain dihydrochloride
Protein synonyms N-(N-Carbonyl-Arg-Val-Arg-al)-Phe dihydrochloride
Environmentally Preferable
Arter
Kilde