ACES ≥99%

Leverandør: Apollo Scientific
Warning

BIA0301-100G BIA0301-250G BIA0301-500G
APOSBIA0301-100GEA 450 NOK
APOSBIA0301-100G APOSBIA0301-250G APOSBIA0301-500G
ACES ≥99%
ACES

An efficient separator in the resolution of proteinsystems by IEF. Biological buffer - useful range 6.4 - 7.4

Formel: H₂NCOCH₂NHCH₂CH₂SO₃H
Smeltepunkt: 270…295 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008030
CAS-nummer: 7365-82-4
EINECS: 230-908-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR