N-(2-Acetamido)-2-aminoethanesulphonic acid (ACES) ≥99%

Supplier: Apollo Scientific
Warning

Synonyms: ACES, N-(Carbamoylmethyl)taurine, N-(Carbamoylmethyl)-2-aminoethanesulfonic acid, N-(2-Acetamido)-2-aminoethanesulphonic acid, N-(Carbamoylmethyl)-2-aminoethanesulphonic Acid, 2-[(Carbamoylmethyl)amino]ethanesulfonic acid

BIA0301-100G BIA0301-250G BIA0301-500G
APOSBIA0301-100GEA 450 NOK
APOSBIA0301-100G APOSBIA0301-250G APOSBIA0301-500G
N-(2-Acetamido)-2-aminoethanesulphonic acid (ACES) ≥99%
N-(2-Acetamido)-2-aminoethanesulphonic acid (ACES)

An efficient separator in the resolution of proteinsystems by IEF. Biological buffer - useful range 6.4 - 7.4

Formula: H₂NCOCH₂NHCH₂CH₂SO₃H
Melting Pt: 270…295 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008030
CAS Number: 7365-82-4
EINECS: 230-908-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR