Alizarin red S (sodium salt)

Leverandør: Apollo Scientific
Warning

BIA0326-100G BIA0326-25G
APOSBIA0326-100GEA 895 NOK
APOSBIA0326-100G APOSBIA0326-25G
Alizarin red S (sodium salt)
Alizarin red S (sodium salt)

C.I. 58005

Formel: C₁₄H₇NaO₇S
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00013049
CAS-nummer: 130-22-3
EINECS: 204-981-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR