ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate])

Leverandør: Apollo Scientific
Warning

BIA1493-10G BIA1493-1G BIA1493-5G
APOSBIA1493-10GEA 5375 NOK
APOSBIA1493-10G APOSBIA1493-1G APOSBIA1493-5G
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate])
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate])

ABTS is a peroxidase substrate used in ELISA procedures.

Formel: C₁₈H₂₄N₆O₆S₄
Smeltepunkt: >300 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00010404
CAS-nummer: 30931-67-0
EINECS: 250-396-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR