Azocarmine G

Leverandør: Apollo Scientific
Warning

Synonymer: Rosinduline, Acid red 101, Basalan Carmine GE

BIA1501-10G BIA1501-25G
APOSBIA1501-10GEA 1800 NOK
APOSBIA1501-10G APOSBIA1501-25G
Azocarmine G
Azocarmine G
Formel: C₂₈H₁₇N₃Na₂O₆S₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00042002
CAS-nummer: 25641-18-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR