N-Acetylglycine(Ac-Gly-OH)

Leverandør: Apollo Scientific
Warning

BIA1745-100G BIA1745-500G
APOSBIA1745-100GEA 340 NOK
APOSBIA1745-100G APOSBIA1745-500G
N-Acetylglycine(Ac-Gly-OH)
N-Acetylglycine(Ac-Gly-OH)
Formel: C₄H₇NO₃
Smeltepunkt: 207…209 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00004275
CAS-nummer: 543-24-8
EINECS: 208-839-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR