EDTA dinatrium salt dihydrat 0,1 M volumetrisk løsning

Leverandør: Apollo Scientific
Warning

BIA181670-1L BIA181670-5L
APOSBIA181670-1LEA 540 NOK
APOSBIA181670-1L APOSBIA181670-5L
EDTA dinatrium salt dihydrat 0,1 M volumetrisk løsning
EDTA dinatrium salt dihydrat
Formel: C₁₀H₁₄N₂Na₂O₈·2H₂O
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00150037
CAS-nummer: 6381-92-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR