4-Bromo-A-23187

Leverandør: Apollo Scientific

Synonymer: Antibiotic A-23187, 4-bromo

BIB1001-1MG BIB1001-5MG
APOSBIB1001-1MGEA 1775 NOK
APOSBIB1001-1MG APOSBIB1001-5MG
4-Bromo-A-23187
4-Bromo-A-23187

Calcium ionophore, analog of calcium ionophore A23187.

Formel: C₂₉H₃₆BrN₃O₆
MW: 602.52 g/mol
Lagringstemperatur: Kjøleskap
CAS-nummer: 76455-48-6

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR